Masterflex® Variable Area Flowmeter Single Tube Frames, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
P02/6C P02/1C P23/6C P22/6C P22/1C P03/6C P03/1C
MFLX03220-08EA 90.38 CAD
MFLX03220-08 MFLX03220-10 MFLX03220-40 MFLX03220-44 MFLX03220-46 MFLX03220-00 MFLX03220-06
Masterflex® Variable Area Flowmeter Single Tube Frames, Avantor®
Flow Meters
Create your own flowmeter systems for complete flexibility.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
  • Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
Choose a frame with a valve port if you plan on adding a valve cartridge to control flow.