Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Panel-Mount Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
ISM4208P ISM4308P ISM4408P ISM4412P
MFLX78001-43EA 4471.4 CAD
MFLX78001-43 MFLX78001-45 MFLX78001-47 MFLX78001-49
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Panel-Mount Peristaltic Pumps, Avantor®
Pumps Custom Engineered Pumps
Independently control flow and direction of each channel via keypad or your PC.
  • Flow rate 0.0002 to 35 ml/min per channel - flow rate depends on drive rpm, rollers, and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel via your computer—expanding your pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensing with the capability of bidirectionalMore Product Information