Masterflex® Mechanically Coupled Centrifugal Pumps, Avantor®
Supplier: VWR International
ACDU200/115T1C ACDU70/315T1C ACDU70/17T3C ACDU70/520T3C ACDU120/530T3C
MFLX70725-00EA 1911.8 CAD
MFLX70725-00 MFLX70725-07 MFLX70725-10 MFLX70725-01 MFLX70725-02 MFLX70725-06 MFLX70725-14
Masterflex® Mechanically Coupled Centrifugal Pumps, Avantor®
Pumps Centrifugal Pumps
Chemical-resistance pump heads provide dependable performance and continuous service.
  • Mechanically close-coupled design saves space and simplifies installation and maintenance
  • Back pullout construction — assembly and overhaul of impeller and seal without distorting suction and discharge connections
  • Top centerline discharge and foot support under casing ensure self-venting and reduce misalignment from pipe loads
The mechanically coupled centrifugal pumps, constructed of 304 stainless steel, are ideal for food and pharmaceutical processing plants, as well as water and wastewater treatment centers. They feature high-quality mechanical shaft seals and O-rings. Also available are optional materials for high-temperature and chemical-duty operations. Contact an Application Specialist for more information.
Delivery information: Includes conduit box for hard-wiring.