Masterflex® L/S® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
7514-10 7514-30 7514-40
MFLX07514-10EA 891.12 CAD
MFLX07514-10 MFLX07514-30 MFLX07514-40
Masterflex® L/S® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Save time with fast tubing changes and reduced maintenance.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Rotor is constructed of stainless steel (SS) for enhanced durability
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® Precision pump tubing sizes
This Easy-Load® pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18.