Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems, Avantor®
Supplier: VWR International
MFLX76000-02 MFLX76000-06 MFLX76000-08
MFLX76000-02EA 3191.6 CAD
MFLX76000-02 MFLX76000-06 MFLX76000-08
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Single-use pump heads are constructed with a disposable polypropylene chamber
  • Reusable pump heads are constructed of durable 316L stainless steel
Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pumpMore Product Information