Masterflex® Six-Channel Syringe Pumps, Avantor®
Supplier: VWR International
MFLX75900-50 MFLX75900-55
MFLX75900-50EA 5040.2 CAD
MFLX75900-50 MFLX75900-55
Masterflex® Six-Channel Syringe Pumps, Avantor®
Pumps Syringe Pumps
Six-channel pump can both withdraw and infuse.
  • Flow rates up to 22.3 ml/min per syringe
  • Network up to 100 pumps from a single communications port on a computer
  • Economical pump can both withdraw and infuse
  • Pumps are capable of holding 41 commands, do unlimited repetitions and unlimited runtime
  • Built-i
Programming commands allow dispensing of a specified amount, automatic rates changes, direction changes, programmed pauses, external or user input, and sending or receive signals from other equipment. Non-volatile memory restores all setup parameters and