Masterflex® Micro Peristaltic Pumps, Avantor®
Supplier: VWR International
MP2-1-PC MP2-2-PC MP2-4-PC
MFLX74906-04EA 5387.8 CAD
MFLX74906-04 MFLX74906-00 MFLX74906-01 MFLX74906-03
Masterflex® Micro Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Operate by computer or on your iOS or Android™ device.
  • Easily achieve flow ranges down to 0.2 µL/min with a 1.26% reproducibility
  • 12-roller head minimizes the pulsation
  • Select pump based upon number of channels needed—from one to eight
  • Wide choice of peristaltic tube sets (0.13 mm i.d. to 3.17 mm i.d.)
Control these micro pumps through your computer using an available USB port and Pump Network Control™ software. Software† can be used remotely on the same Wi-Fi network using an Apple® iOS or Android device. The Pump Network Control software controls dire