Masterflex® L/S® Easy-Load® II Pump Head Tubing Retainer Kits, Avantor®
Supplier: Avantor Fluid Handling
77200-90 77200-92 77200-94
MFLX77200-90EA 130.19 CAD
MFLX77200-90 MFLX77200-92 MFLX77200-94
Masterflex® L/S® Easy-Load® II Pump Head Tubing Retainer Kits, Avantor®
Pumps Peristaltic Pumps
Enhance the life of your pump head with the replacement retainer kits.
  • Retainer kits provide the parts necessary to replace the spring-loaded tubing retainers in your Masterflex® Easy-Load® II pump heads
Replacement parts to keep your pump head operating even longer. Tubing retainers hold the tubing in place within the pump head. Pump head sold separately.
Delivery information: Includes two small upper retainers, two small lower retainers, and two compression springs.