Masterflex® Drum Pump ODP Motors, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
SP-280P
MFLX70604-00EA 1448.86 CAD
MFLX70604-00
Masterflex® Drum Pump ODP Motors, Avantor®
Pumps Drum/Transfer Pumps
Safety transfer a variety of liquids from drums, tanks, and small storage vessels.
  • Maintenance is reduced with no mechanical seals
  • Run dry capability
This drum pump motor is designed to transfer a variety of materials from 55 gallon drums and tanks. Drum pump tubes are sold separatley, making this drum pump customizable to your specific application. Pump tubes are available in multiple materials and lengths, model numbers 70604-42, -44, -52, -54, -62, -64, -66, and -68. Barrel adapter 70604-92 sold separately.
The motor has thermal overload protection which stops the pump in case of overload, allowing it to cool down.