Masterflex® Plastic Ball Valves, Two-way Straight, PVC, Avantor®
Supplier: SPECIALTY MFG CO
PVC226-2F2F-E PVC226-2F2M-F PVC226-4F4F-E PVC226-4F4F-F PVC226-4F4M-E PVC226-4F4M-F PVC226-4B4B-E PVC226-4B4B-F PVC226-6B6B-F PVC226-8B8B-E PVC-638-4F4F-E PVC-638-4F4M-E PVC-638-4B4B-E PVC-638-TB4F-B PVC-638-4FTB-E PVC-638-4MTB-B PVC-638-4MTB-E PVC-638-4CJ4CJ-E PVC-638-4M4CJ-B PVC-638-4M4CJG-B PVC-638-6F6F-E PVC-638-6F6M-B PVC-638-6M6F-E PVC-638-6B6B-E PVC-638-6F6CJG-E PVC-638-6M6CJG-B PVC 226-4B4B-B PVC 226-4SMC4SMC-B PVC-638-4F4F-B PVC-638-4B4B-B PVC226-2F2F-F PVC226-2F2M-E PVC 226-4SMC4SMC-E PVC226-4SMC4SMC-F PVC226-6B6B-E PVC 226-8B8B-F PVC-638-4CJ4CJ-B PVC-638-4F4CJ-B PVC-638-4F4CJ-E PVC-638-4M4CJ-E PVC-638-4SMC4SMC-E PVC-638-4F4CJG-B PVC-638-4F4CJG-E PVC-638-4M4CJG-E PVC-638-6F6F-B PVC-638-6FTB-B PVC-638-6FTB-E PVC-638-6MTB-B PVC-638-6MTB-E PVC-638-6CJ6CJ-B PVC-638-6CJ6CJ-E PVC-638-6F6CJG-B PVC-638-6M6CJG-E PVC 226-2F2F-B PVC 226-6CJG6CJG-B PVC 226-8B8B-B PVC-638-6B6B-B PVC-638-6CJG6CJG-B PVC-638-6CJG6CJG-E
MFLX01377-01EA 39.66 CAD
MFLX01377-01 MFLX01377-10 MFLX01377-16 MFLX01377-18 MFLX01377-24 MFLX01377-26 MFLX01377-30 MFLX01377-32 MFLX01377-44 MFLX01377-50 MFLX01377-80 MFLX01377-82 MFLX01377-84 MFLX01377-86 MFLX01377-88 MFLX01377-90 MFLX01377-92 MFLX01377-96 MFLX01378-03 MFLX01378-12 MFLX01378-18 MFLX01378-20 MFLX01378-22 MFLX01378-24 MFLX01378-40 MFLX01378-42 MFLX01378-87 MFLX01378-89 MFLX01379-08 MFLX01379-10 MFLX01377-03 MFLX01377-08 MFLX01377-38 MFLX01377-40 MFLX01377-42 MFLX01377-52 MFLX01377-94 MFLX01377-98 MFLX01378-01 MFLX01378-04 MFLX01378-06 MFLX01378-08 MFLX01378-10 MFLX01378-14 MFLX01378-16 MFLX01378-26 MFLX01378-28 MFLX01378-30 MFLX01378-32 MFLX01378-34 MFLX01378-36 MFLX01378-38 MFLX01378-44 MFLX01378-78 MFLX01378-92 MFLX01378-95 MFLX01379-12 MFLX01379-13 MFLX01379-14
Masterflex® Plastic Ball Valves, Two-way Straight, PVC, Avantor®
Valves Ball Valves
Nonmetallic wetted parts ideal for water and mild aqueous solutions.
  • Maximum pressure: 125 psi (8.6 bar)
  • Maximum temperature: 140 °F (60 °C)
Ball valves are designed primarily for on/off service.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.