Masterflex® ACR Valves for Chemical and Corrosive Fluids, PTFE, Avantor®
Supplier: MIDWEST VALVE & FITTING INC
CR-PVC-2W4F-C MFLX98516-02 MFLX98516-10 MFLX98516-12
MFLX98516-18EA 1077.56 CAD
MFLX98516-18 MFLX98516-02 MFLX98516-10 MFLX98516-12
Masterflex® ACR Valves for Chemical and Corrosive Fluids, PTFE, Avantor®
Valves Solenoid Valves
Chemical-resistant valves provide precise fluid control.
  • Manufactured in a clean room environment
  • Process connections of NPT(F), compression, and hose barb
  • Long service life performing in the harshest of applications
These pneumatically operated valves feature chemically resistant wetted surfaces constructed from PTFE—the material of choice for both thermal resistance and chemically inert applications, or many difficult-to-handle media. Commonly used in wet benches and process work stations, these valves are ideal for DI water and other harshMore Product Information