Masterflex® Particle Tolerant Solenoid Valves, Avantor®
Supplier: CLARK SOLUTIONS
MTV-2K-N1/4UG 24V CTV-3-1/4U-K-G-24V MTV-3K-1/4U-G-24V MFLX98305-24 MFLX98305-32 MFLX98305-36
MFLX98305-26EA 282.82 CAD
MFLX98305-26 MFLX98305-34 MFLX98305-42 MFLX98305-24 MFLX98305-32 MFLX98305-36
Masterflex® Particle Tolerant Solenoid Valves, Avantor®
Valves Solenoid Valves
Particles will not cause damage.
  • Special elastomer diaphragm and seals are forgiving of particulate matter that would typically damage a plastic valve seat
  • Inert materials — excellent chemical resistance
  • Molded PEEK valve bodies
  • Excellent choice for demanding low-flow applications where minimum size and internal volume are important
These Masterflex® particle tolerant solenoid valves feature a apecial Perfluor™ soft elastomer diaphragm and seals, which are forgiving of particulate matter that would typically damage a plastic valve seat. Valves are constructed with a PEEK body, PTFE diaphragm, Perfluor seat and seal, and ceramic stem. Choose from two-way normally closed valves, two-way normally open valves, and three-way valve options.