Masterflex® Stepping Motor Proportioning Valves, Avantor®
Supplier: VWR International
SMV30-A SMV40-A MFLX98651-00 MFLX98651-06 MFLX98651-10
MFLX98651-02EA 1543.66 CAD
MFLX98651-02 MFLX98651-04 MFLX98651-08 MFLX98651-00 MFLX98651-06 MFLX98651-10
Masterflex® Stepping Motor Proportioning Valves, Avantor®
Valves Solenoid Valves
Resist pressures up to 500 psi!
  • High precision, two-way metering valves in aluminum or 316 SS for air/water
  • Operate continuously without coil overheating problems found in traditional solenoid designs
Stainless steel models include a 316 SS valve blocks, PTFE-lined aluminum housing blocks, Viton® O-rings, and PFA closing pins.
Aluminum models include aluminum housings and valve blocks, Viton® O-rings, and PFA closing pins.
Also includes a 9-pin "D" electrical connector. Optional connection cable (98651-50) for easy input/output wiring. Power input is +12 VDC at 800 mA, protected by a resettable fuse.