Masterflex® Check Valves, PTFE, Avantor®
Supplier: MIDWEST VALVE & FITTING INC
CV-4-4-P-05 CV-6-6-P-05 CV-8-8-P-05 CV-2-2-P-05
MFLX98515-14EA 752.08 CAD
MFLX98515-14 MFLX98515-16 MFLX98515-18 MFLX98515-12
Masterflex® Check Valves, PTFE, Avantor®
Valves Check Valves
Use with caustic and high-purity liquids in many applications.
  • Constructed of durable PTFE
Designed for use with corrosive fluids of varying viscosities in chemical, microelectronics, and petroleum industries.