Masterflex® Transfer Tubing Spool, Polytetrafluoroethylene (PTFE), Avantor®
Supplier: VWR International
MFLX95231-00 MFLX95231-08 MFLX95231-10 MFLX95231-12 MFLX95231-14 MFLX95231-02 MFLX95231-06
MFLX95231-00EA 1545.24 CAD
MFLX95231-00 MFLX95231-08 MFLX95231-10 MFLX95231-12 MFLX95231-14 MFLX95231-02 MFLX95231-06
Masterflex® Transfer Tubing Spool, Polytetrafluoroethylene (PTFE), Avantor®
Tubing Transfer Tubing
Available in spooled rolls for increased versatility.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.