Masterflex® Transfer Tubing, Terminator®, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
TERMINATOR,2",25FT TERMINATOR,1.5",25FT TERMINATOR,1",50FT TERMINATOR,1/4",100FT TERMINATOR,3/4",50FT TERMINATOR,1/2",100FT
MFLX95954-11EA 1008.04 CAD
MFLX95954-11 MFLX95954-01 MFLX95954-03 MFLX95954-05 MFLX95954-07 MFLX95954-09
Masterflex® Transfer Tubing, Terminator®, Avantor®
Tubing Transfer Tubing
Superior abrasion resistance at high pressure.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Outstanding resistance to heat and ozone
Masterflex® Terminator® transfer tubing is ideal for use with severe service applications due to its superior abrasion resistance. The Buna-N nitrile tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube and tube cover meet RMA Class A high oil resistance and nonconductive at 1000 VDC certification.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.