Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Chem-Durance® Bio, Avantor®
Supplier: VWR International
MFLX96328-14 MFLX96328-18 MFLX96328-24 MFLX96328-26 MFLX96328-30 MFLX96328-34 MFLX96328-36 MFLX96328-42 MFLX96328-48
MFLX96328-14EA 241.74 CAD
MFLX96328-14 MFLX96328-18 MFLX96328-24 MFLX96328-26 MFLX96328-30 MFLX96328-34 MFLX96328-36 MFLX96328-42 MFLX96328-48
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Chem-Durance® Bio, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Long service life - Outstanding fatigue resistance due to flexing!
  • Suitable for applications using acetone, methyl-ethyl-ketone (MEK), and other highly aggressive chemicals
To complete yourMore Product Information