Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® Fuel and Lubricant, Avantor®
Supplier: VWR International
MFLX95703-18 MFLX95703-32 MFLX95703-14 MFLX95703-22 MFLX95703-24 MFLX95703-26 MFLX95703-48 MFLX95703-36 MFLX95703-38 MFLX95703-44 MFLX95703-12 MFLX95703-42 MFLX95703-10 MFLX95703-34 MFLX95703-40 MFLX95703-30
MFLX95703-18EA 265.44 CAD
MFLX95703-18 MFLX95703-32 MFLX95703-14 MFLX95703-22 MFLX95703-24 MFLX95703-26 MFLX95703-48 MFLX95703-36 MFLX95703-38 MFLX95703-44 MFLX95703-12 MFLX95703-42 MFLX95703-10 MFLX95703-34 MFLX95703-40 MFLX95703-30
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® Fuel and Lubricant, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Specially designed to transport hydrocarbons, petroleum products, or distallates
  • Transparent yellow color
Appearance: Opaque Temperature: -70 to 52 °C Life at 100 rpm: 1000 hrs Advantages:Long service life, outstanding fatigue resistance due to flexing Application:Suitable for applications using acetone, methyl-ethyl-ketone (MEK), and other highly aggressive chemicals The overall length of Tygon Fuel & Lubricant F-4040-A 95715-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long. Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.