Masterflex® Ismatec® Microbore Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
FA0586 FA0112 F09656 F09658 F09597
MFLX95590-18EA 466.1 CAD
MFLX95590-18 MFLX95590-26 MFLX95590-30 MFLX95590-34 MFLX95590-42 MFLX95590-48
Masterflex® Ismatec® Microbore Pump Tubing, Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Smooth surface and low protein binding levels.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Use platinum-cured silicone tubing for high purity applications where outgassing of peroxide product is not allowed.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.