Masterflex® Low-Flow Peristaltic Pump Replacement Tubing Sets, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
SP101.980CP SP102.980CP SP100.980CP
MFLX73160-58EA 83.74 CAD
MFLX73160-58 MFLX73160-56 MFLX73160-54
Masterflex® Low-Flow Peristaltic Pump Replacement Tubing Sets, Avantor®
Tubing Peristaltic Pump Tubing
Choose tubing based on chemical compatibility, flow rate, and your current pump.
  • Global market leader in range of formulations for most extensive chemical compatibility
Choose from silicone, norprene and viton replacement tubing for your peristaltic pump.