Masterflex® I/P® Precision Pump Tubing, Tygon® E-Lab, Avantor®
Supplier: VWR International
MFLX06509-26 MFLX06509-73 MFLX06509-82
MFLX06509-26EA 616.2 CAD
MFLX06509-26 MFLX06509-73 MFLX06509-82
Masterflex® I/P® Precision Pump Tubing, Tygon® E-Lab, Avantor®
Tubing Peristaltic Pump Tubing
No BPA or phthalates—environmentally responsible, more sustainable, and safer.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
Masterflex® Tygon® E-Lab PrecisionMore Product Information