Masterflex® I/P® High-Performance Precision Pump Tubing, Tygon® E-Lab, Avantor®
Supplier: VWR International
MFLX06509-70 MFLX06509-88 MFLX06509-89
MFLX06509-70EA 1039.64 CAD
MFLX06509-70 MFLX06509-88 MFLX06509-89
Masterflex® I/P® High-Performance Precision Pump Tubing, Tygon® E-Lab, Avantor®
Tubing Peristaltic Pump Tubing
No BPA or phthalates—environmentally responsible, more sustainable, and safer.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
Masterflex® Tygon® E-Lab High-Performance Precision pump tubing is specially formulated for resistance to flex-fatigue and abrasion and offers superior life, which minimizes the labor and expense of replacement. Its clear, glassy smooth inner bore helps prevent buildup to facilitate cleaning and tubing transparency makes for easy flow monitoring.
Ideal for general laboratory, food and beverage, and biopharma applications, this tubing complies with USP Class VI, FDA, EU Foods, and NSF standards.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.