Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® E-Lab, Avantor®
Supplier: VWR International
MFLX06509-15 MFLX06509-24 MFLX06509-35 MFLX06509-36
MFLX06509-15EA 364.98 CAD
MFLX06509-15 MFLX06509-24 MFLX06509-35 MFLX06509-36
Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® E-Lab, Avantor®
Tubing Peristaltic Pump Tubing
Ideal for general transfer applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: General laboratory use
Characteristics: Nontoxic, nonaging, nonoxidizing, easy flow monitoring, transparent
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.