Masterflex® L/S® Tube Sets, PTFE, Avantor®
Supplier: Avantor Fluid Handling
77390-50 77390-60
MFLX77390-50CS 0 CAD
MFLX77390-50 MFLX77390-60
Masterflex® L/S® Tube Sets, PTFE, Avantor®
Tubing Peristaltic Pump Tubing
Excellent chemical resistance and chemically inert.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
For use with Masterflex® L/S® PTFE Tubing Pump Head
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.