Masterflex® L/S® 2-Stop Pump Tubing, Chem-Durance® Bio, Avantor®
Supplier: VWR International
MFLX96117-13 MFLX96117-14 MFLX96117-15 MFLX96117-16 MFLX96117-25 MFLX96117-35
MFLX96117-13EA 300.2 CAD
MFLX96117-13 MFLX96117-14 MFLX96117-15 MFLX96117-16 MFLX96117-25 MFLX96117-35
Masterflex® L/S® 2-Stop Pump Tubing, Chem-Durance® Bio, Avantor®
Tubing Peristaltic Pump Tubing
Excellent chemical resistance and long pumping life.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Laboratory, pharmaceutical processing, and biotech
Characteristics: Excellent chemical resistance, excellent life and durability under pressure, low spallation, plasticizer-free inner liner with elastomer jacket, opaque
For use with Masterflex® multichannel pump heads.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.