Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, Avantor®
Supplier: VWR International
MFLX06508-13 MFLX06508-16 MFLX06508-17 MFLX06508-18 MFLX06508-25 MFLX06508-14
MFLX06508-13CS 0 CAD
MFLX06508-13 MFLX06508-16 MFLX06508-17 MFLX06508-18 MFLX06508-25 MFLX06508-14
Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
Protect your light-sensitive samples.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.