Masterflex® Reducer Fittings, Hose Barb, Straight (PVDF), Avantor®
Supplier: VWR International
MFLX40614-52 MFLX40614-55 MFLX40614-57 MFLX40614-58 MFLX40614-60 MFLX40614-61 MFLX40614-62 MFLX40614-64 MFLX40614-65 MFLX40614-66 MFLX40614-68 MFLX40614-69 MFLX40614-70 MFLX40614-71 MFLX40614-72 MFLX40614-73 MFLX40614-74 MFLX40614-75 MFLX40614-76 MFLX40614-77 MFLX40614-78 MFLX40614-63 MFLX30703-58 MFLX40614-53 MFLX40614-56 MFLX40614-67 MFLX40614-59 MFLX40614-51
MFLX40614-52EA 22.59 CAD
MFLX40614-52 MFLX40614-55 MFLX40614-57 MFLX40614-58 MFLX40614-60 MFLX40614-61 MFLX40614-62 MFLX40614-64 MFLX40614-65 MFLX40614-66 MFLX40614-68 MFLX40614-69 MFLX40614-70 MFLX40614-71 MFLX40614-72 MFLX40614-73 MFLX40614-74 MFLX40614-75 MFLX40614-76 MFLX40614-77 MFLX40614-78 MFLX40614-63 MFLX30703-58 MFLX40614-53 MFLX40614-56 MFLX40614-67 MFLX40614-59 MFLX40614-51
Masterflex® Reducer Fittings, Hose Barb, Straight (PVDF), Avantor®
Fittings and Connectors
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range -80 to 265 °F (-62 to 129 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.