Masterflex® Concentric Reducer Fittings, Sanitary Tri-Clamp, Straight, Avantor®
Supplier: VWR International
MFLX31200-32 MFLX31200-34 MFLX31200-35 MFLX40618-17 MFLX40618-15 MFLX40618-16
MFLX31200-32EA 155.79 CAD
MFLX31200-32 MFLX31200-34 MFLX31200-35 MFLX40618-17 MFLX40618-15 MFLX40618-16
Masterflex® Concentric Reducer Fittings, Sanitary Tri-Clamp, Straight, Avantor®
Fittings and Connectors
Maintain sanitary conditions.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Surface finishes to 15 micron minimizes adhesion or entrapment
  • Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
  • Compatible with other Tri-Clamp® connections
The sanitary Tri-Clamp concentric reducers are ideal for use in the biopharmaceutical, food and beverage, and single-use disposable industries. The fittings are made from animal-derived ingredient-free and phthalate-free polypropylene (PP) and PVDF. They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave. The PVDF fittings withstand higher temperatures than the PP fittings and offer extremely low surface tension for lower protein and lipid binding.