Nitrite, Titrets Titration Cells, CHEMetrics
Supplier: CHEMetrics
K-7025 K-7050
CXK-7025EA 110.93 CAD
CXK-7025 CXK-7050
Nitrite, Titrets Titration Cells, CHEMetrics
Environmental Test Kits
Ceric sulfate is the titrant and ferroin is the end point indicator. The method is free from glycol interference in samples that contain up to 75% glycol, making it particularly applicable to systems that contain nitrite corrosion inhibitors. Results are expressed as ppm (mg/L) NaNO2.
  • Dependable, sealed single-use titrants
  • Fast, easy procedure — no need to count drops
  • No burettes or other equipment necessary
  • Perform a titration anywhere
Titrets ampoules use reverse titration to quantify concentrations. After snapping the ampoule tip, the sample is drawn into the ampoule in small doses (with the Titrettor device included in each kit that precisely controls the sample) (Step 1), until a color change signals that the equivalence point has been reached (Step 2). The titration is stopped at the end point and the ampoule is held upright. The liquid level will correspond to a printed scale on the ampoule's outer surface (Step 3).
Nitrite, an intermediate in the nitrogen cycle, is formed during the decomposition of organic matter but readily oxidizes to form nitrate. These processes occur in wastewater treatment plants, water distribution systems, and natural waters. Nitrites are useful as corrosion inhibitors, preservatives, pigments, and in manufacturing many organic preservative chemicals. A Maximum Contaminant Level of 1 mg/L has been established by the USEPA for nitrite-nitrogen in drinking water.
K-7025 Increments: 250, 275, 300, 325, 350, 375, 400, 450, 500, 625, 750, 875, 1000, 1250, 1750, 2500 ppm
K-7050 Increments: 500, 550, 600, 650, 700, 750, 800, 900, 1000, 1250, 1500, 1750, 2000, 2500, 3500, 5000 ppm
Delivery information: Kits come in a cardboard box and contain everything needed to perform 30 Nitrite titration tests: thirty ampoules with valve assemblies, titrettor, 25 mL sample cup, and instructions.