Cole-Parmer® SH-200 Series Gyro Rocking Shakers, Antylia Scientific
Supplier: Antyila Scientific
51901-05 51901-13
76542-142EA 2894.73 CAD
76542-142 76542-144
Cole-Parmer® SH-200 Series Gyro Rocking Shakers, Antylia Scientific
Shakers Rotating/Rocking Shakers
Gentle swirling motion prevents foaming.
  • Adjustable tilt angle for optimal mixing
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be used in incubators
These gyro rocking shakers feature a 3D gyrating action with a 3 to 12 degree tilt angle. You can easily adjust the angle of the tilt by hand for optimal performance. Best suited for low-foaming requirements, the shakers are ideal for cell culturing, staining and de-staining. The shakers offer a quiet drive mechanism, easy to use controller with timer, LED display and a digital speed selection from 5 to 70 rpm. Platform lip and non-slip mat hold vessels securely.
Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert. Shaker is capable of continuous use and the microprocessor maintains speed accuracy over long runs.
Ordering information: Supplied with flat platform, non-slip mat, and power cord.