VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
76518-334 76518-336 76518-338 76518-340 76518-342
76518-334CS 319.82 CAD
76518-334 76518-336 76518-338 76518-340 76518-342
VWR® Nitrile Examination Gloves, Blue
Gloves
Ambidextrous gloves are soft and comfortable for everyday use. They protect hands from a broad range of chemicals. Beaded cuff allows easy donning.
  • Finger textured
  • Ambidextrous
  • Nonsterile
  • Soft and comfortable
  • Protects hands from broad range of chemicals and hazards
  • Powder-free
Ideal for use in industrial and laboratory applications.
Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverseMore Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Material Nitrile
Finish Textured fingers
Cuff style Beaded
Sterility Non sterile