LyoPrime Luna™ Probe One-Step RT-qPCR Mix with UDG
Supplier: New England Biolabs (NEB)
L4001S L4001P
CA76540-506EA 353 CAD
CA76540-506 CA77322-316
LyoPrime Luna™ Probe One-Step RT-qPCR Mix with UDG
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
The LyoPrime Luna™ Probe One-Step RT-qPCR Mix with UDG is offered in a lyophilized format, allowing for ambient/room temperature shipping and storage prior to use.
  • Ambient/room temperature shipping and storage; two-year shelf life at room temperature
  • Maximize your throughput by multiplexing up to five targets
  • Reduce risk of carryover contamination with thermolabile UDG and dUTP included
  • Unique passive reference dye offers compatibility across a variety of instrument platforms
  • Contains non-fluorescent blue tracking dye for ease of pipetting
The mix contains all necessary components for one-step RT-qPCR except primers, probe(s), and RNA sample. It can be reconstituted by simply adding water to a 2 to 4X concentration, allowing for increased sample input. Performance in multiplexing applications has been optimized, with sensitive, linear detection achieved for up to five targets across a range of inputs.
Packaging: Available in 2×60 reaction glass vials or a 96-well plate (one reaction per well).