Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.8 µm, Whatman Products (Cytiva)
Supplier: Cytiva
10408915 10409970 10408970 10408915 10408970 10409970
CA11012-796PK 1890.32 CAD
CA11012-796 CA11012-906 CA11013-036 10034-974 10034-976 10034-988
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.8 µm, Whatman Products (Cytiva)
Filters Membrane Filters
Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
  • Sterile options available for critical applications
  • Excellent contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate
  • The membrane offers a high degree of internal surface area for greater adsorption
More Product Information