Whatman™ Mixed Cellulose Ester Membranes, Non-Sterile, 0.45 µm
Supplier: Cytiva
7141-004 7141-204 10406812 10401606 10401612 10401614 10401618 10401606 10401614 10401612 10401618 10401631 10409714 10406814 10401621 10401626 10409414 10406512 10401621 10401626 10401631 10406512 10406814 10409414 10409714 10406812 7141-004 7141-204 10401664
28297-730PK 364.74 CAD
28297-730 28297-738 89187-132 CA74300-040 CA74300-042 CA74330-192 CA11008-578 74300-040 74330-192 74300-042 11008-578 CA11012-736 CA11013-040 CA11013-472 CA11013-578 CA11013-580 CA11013-970 CA11013-972 10034-910 10034-912 10034-914 10034-958 10034-962 10035-796 10035-798 CA89187-132 CA28297-730 CA28297-738 CA76423-744
Whatman™ Mixed Cellulose Ester Membranes, Non-Sterile, 0.45 µm
Filters Membrane Filters
Mixed cellulose ester, WME range, circles, gridded. Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
  • Sterile options available for critical applications
  • Excellent contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate
  • The membrane offers a high
More Product Information