Water, HyClone™ HyPure USP, WFI (water for injection) quality

Supplier: Cytiva

SH31191.01 SH31191.02 SH31191.03 SH31191.LS SH31191.04
CA76538-678EA 43.05 CAD
CA76538-678 CA76538-680 CA76538-682 CA76538-676 CA76894-126
Water, HyClone™ HyPure USP, WFI (water for injection) quality
Water

Support cell culture growth and biomanufacturing processes with Water for Injection (WFI) quality water.


  • Meets stringent USP specifications
  • Manufactured in ISO 9001 certified facility
  • 0.1 µm sterile filtered
  • Shelf life 24 months


HyClone HyPure WFI Quality Water undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP). Water for injection (WFI) is produced in ISO 9001 certified facilities to ensure a high quality product.


HyPure WFI Quality Water meets the following criteria: Must be produced from a validated water purification system that meets or exceeds current USP requirements for HyPure WFI Quality Water. Must have a quality assurance monitoring system that ensures continuous compliance and incorporates system test and action limits. Must meet all applicable chemical, purity, and other acceptance criteria as outlined in the current USP for both system and packaged water. Must include a lot-specific CoA for each lot of water produced, listing both acceptance criteria and actual test results.


Caution: Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.

Formula: H₂O
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR