Muffle Furnaces up to 1100 °C, Models L and LT, Nabertherm
Supplier: NABERTHERM INC.
L-034H1LN1-120 L-034H1LN1-208 L-034H1LN1-240 L-034K1LN1-120 L-034K1LN1-208 L-034K1LN1-240 L-054H1LN1-208 L-054H1LN1-240 L-054K1LN1-208 L-054K1LN1-240 L-094H1LN1-208 L-094H1LN1-240 L-094K1LN1-208 L-094K1LN1-240 L-154H1LN1-208 L-154H1LN1-240 L-154K1LN1-208 L-154K1LN1-240 L-244H1LN1-480 L-244K1LN1-480 L-404H1LN1-480 L-404K1LN1-480 L-404H1LN1-208 L-404K1LN1-208
76537-736EA 7969.68 CAD
76537-736 76537-738 76537-740 76537-742 76537-744 76537-746 76537-748 76537-750 76537-752 76537-754 76537-756 76537-758 76537-760 76537-762 76537-764 76537-766 76537-768 76537-770 76537-772 76537-774 76537-776 76537-778 76537-780 76537-782 CA76639-010 CA76639-014
Muffle Furnaces up to 1100 °C, Models L and LT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These muffle furnaces have been proven for daily laboratory use. These models stand out for their workmanship, advanced and attractive design and high level of reliability. The muffle furnaces come equipped with either a fold down or lift up door at no extra charge.
  • Tmax 1100 °C
  • Heating from two sides by ceramic heating plates (heating from three sides for muffle furnaces L 24/11 - LT 40/12)
  • Thermocouple type S; manual or automatic gas supply system
  • Ceramic heating plates with integral heating element which is safeguarded and easy to replace
  • Optional fold
More Product Information