Water, meetSpec_Ph.Eur.,USP injection_WFI, Cell Culture, sterile, Corning®

Supplier: Corning

Synonyms: Hydrogen oxide

25-065-LB 25-065-LH 25-065-LG
CA95029-816EA 973.92 CAD
CA95029-816 CA95029-820 CA95029-818
Water, meetSpec_Ph.Eur.,USP injection_WFI, Cell Culture, sterile, Corning®
Water

Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).


Technical Advantage: Wide range of applications in processes requiring very high levels of sterility USP/EP Certified Water is validated to the most recent specifications found in the United States Pharmacopeia (USP) and the European Pharmacopeia (EP). The specific testing is in accordance to testing for "Sterilized Water for Injection", (European Pharmacopeia), and "Sterile Water for Injection", (United States Pharmacopeia).

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

Specification Test Results

Mycoplasma Tested Negative
LM (plating effciency) Pass
Oxidizable Substances Pass
Total Organic Carbon (before packaging) USP <500 ppb
Water Conductivity (packaged water) USP Meets USP requirements
Water Conductivity (before packaging) USP Meets USP Temperature/Conductivity Requirements
Endotoxin USP ≤0.025 EU/ml
Sterility USP Pass
Acidity-Alkalinity Tested in Accordance with EP
Chlorides Pass
Nitrates ≤0.2 ppm
Sulphates Pass
Ammonium ≤0.2 ppm
Calcium and Magnesium Pass
Residue on Evaporation ≤3 mg (0.003)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR