VWR® Economy Specimen Containers, PP
Supplier: VWR International
76532-294 76532-296 76532-298 76532-300 76532-302 76532-304
76532-294CS 177.02 CAD
76532-294 76532-296 76532-298 76532-300 76532-302 76532-304
VWR® Economy Specimen Containers, PP
Sample Containers General Purpose Containers
Sturdy, shatter-resistant containers are manufactured from transparent polypropylene to allow constant sample viewing.
  • Sturdy, shatter-resistant design
  • Transparent siding
  • Polypropylene construction
  • Available in nonsterile or sterile formats
  • Containers feature molded graduations in 10 mL and 0.5 oz. increments
  • Polyethylene snap-on lids are sold separately, except where noted
Lids create a secure seal, and offer space for date, name, doctor, and room number molded into the surface. Lids fit both 5 and 8 oz. containers.
Certifications: Manufactured under EN ISO13485:2016.