High-Temperature Tube Furnaces with SiC Rod Heating up to 1500 °C, RHTC Series, Nabertherm
Supplier: NABERTHERM INC.
RC182L5A-480 RC182O5A-480 RC282L5A-480 RC282O5A-480 RC382L5A-480 RC382O5A-480
76538-046EA 23414.68 CAD
76538-046 76538-048 76538-050 76538-052 76538-054 76538-056
High-Temperature Tube Furnaces with SiC Rod Heating up to 1500 °C, RHTC Series, Nabertherm
Ovens Tube Furnaces
The RHTC 80/230/15, RHTC 80/450/15, RHTC 80/710/15 high-temperature tube furnaces are heated by SiC heating elements which are arranged parallel to the working tube which provides excellent temperature uniformity.
  • Tmax 1500 °C
  • Represent an inexpensive variant in the high-temperature range
  • SiC heating elements are easy to replace
  • Type S thermocouple
  • Active cooling of housing for low surface temperatures
  • Programable: 5 programs, 4 segments
Working tube contains charge control with temperature measurement. Over-temperature limiter with adjustable cutoutMore Product Information