VWR® Sterile Standard Hexagonal Weighing Boats, White
Supplier: VWR International
76540-492 76540-494
76540-492CS 586.02 CAD
76540-492 76540-494
VWR® Sterile Standard Hexagonal Weighing Boats, White
Boats Weighing Boats
These hexagonal weighing boats are flexible and feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
  • Electron beam sterilized to reduce the risk of cross-contamination and compromised samples
  • Made from 100% virgin anti-static polystyrene that is waterproof and reduces the chance of materials sticking to the units
  • Disposable
  • Flat bottoms and sloping sides provide accurate transfer
  • Can be used with microwave moisture analyzers
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases
  • Color:
More Product Information