Translucent Snap Caps, Globe Scientific
Supplier: Globe Scientific
113144G 113144R 113144Y 113144B 113144L 113144C 113144A 113143Y 113143R 113143B 113143L 113143C 113143G 113143A
76538-862PK 64.74 CAD
76538-862 76538-864 76538-866 76538-868 76538-870 76538-872 76538-874 76538-876 76538-878 76538-880 76538-882 76538-884 76538-886 76538-888
Translucent Snap Caps, Globe Scientific
Closures Test/Sample Tube Closures
These colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes and plastic culture tubes.
  • Economical way to recap glass tubes, regardless of manufacturer
  • Available in seven (7) translucent colors that correspond to Vacutainer® cap colors
  • Caps can be written on with indelible ink for easy sample identification
  • Prevents spills and evaporation by employing a liquid tight seal
These caps are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation and refrigeration procedures. They are available in bright translucent colors for sample identification purposes. The caps feature a single thumb tab for easy removal and close nesting in tube racks.