Vacuum Tubing, Tygon, Ace Glass
Supplier: Ace Glass
12696-09 12696-11
76529-702EA 1325.34 CAD
76529-702 76529-704
Vacuum Tubing, Tygon, Ace Glass
Tubing Transfer Tubing
Tygon S3™ E-3603 phthalate-free vacuum tubing will withstand full vacuum at room temperature.
  • Resists most inoraganic chemicals
  • Can be used in corrosive atmospheres
Withstands full vacuum at room temperature (29.9" [760 mm] Hg at 73 °F [23 °C] and up to 27" [686 mm] Hg at 140 °F [60 °C]).
Delivery information: Material complies with FDA NSF 3-A and meets USP class VI.