Vacuum Tubing, Flexible Stainless Steel, Ace Glass
Supplier: Ace Glass
12694-16 12694-25 12694-40
76529-696EA 1279.19 CAD
76529-696 76529-698 76529-700
Vacuum Tubing, Flexible Stainless Steel, Ace Glass
Tubing Transfer Tubing
Flexible stainless steel accordion vacuum tubing featuring welded NW (DN) flanges.
  • Welded NW flanges
  • Glass to metal connection
Connect a NW glass flange to a vacuum pump.
Delivery information: Includes silicone O-Rings.