Filter Reaction Vessels, Jacketed, Ace Glass
Supplier: Ace Glass
12650-10 12650-20 12650-30 12650-50 12650-75 12650-100 12650-150
76529-682EA 11854.72 CAD
76529-682 76529-684 76529-686 76529-688 76529-690 76529-692 76529-694
Filter Reaction Vessels, Jacketed, Ace Glass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Kilo scale, cylindrical glass vessels are flanged on both ends.
  • Unique jacketed flange delivers cooling/heating capacity in what is for other manufacturers a 'dead zone'
  • Consistant interior bore-thru diameter from top to bottom
  • Temperature range is –60 to +200 °C
  • Maximum jacket pressure is 8 psig
Various size vessels are available, ranging from 10 L through 150 L.