Metal to Metal Connection, NW Flange, Ace Glass
Supplier: Ace Glass
12198-10 12198-14 12198-25 12198-26 12198-29 12198-40 12198-41 12198-44
76529-662EA 31.88 CAD
76529-662 76529-666 76529-668 76529-670 76529-672 76529-674 76529-676 76529-678 76529-664
Metal to Metal Connection, NW Flange, Ace Glass
Seals Gaskets
Connections for NW metal flanges to like NW metal flanges.
  • Connect vacuum pump to stainless steel tubing
  • Various clamp and gasket options available
Included Silicone O-ring is suitable for use in temperatures from –60 to +205 °C. Viton O-ring is suitable for use in temperatures from –26 to +240 °C.
Delivery information: Gasket and clamp sold separately.