Metal to Glass Gasket, NW Flange, Ace Glass
Supplier: Ace Glass
12197-02 12197-04 12197-06 12197-32 12197-34 12197-36
76529-650EA 332.85 CAD
76529-650 76529-652 76529-654 76529-656 76529-658 76529-660
Metal to Glass Gasket, NW Flange, Ace Glass
Seals Gaskets
NW (DN) flange metal to glass sealing gasket featuring a PTFE carrier and O-ring.
  • High temperature metal to glass seal
  • Made of PTFE
  • Excellent for vaccum work or a maximum of 2 bar pressure
Included Capfe O-ring is suitable for use in temperatures from –60 to +204 °C. Viton O-Ring is suitable for use in temperatures from –26 to +240 °C.