Glass to Glass Gasket, NW Flange, Ace Glass
Supplier: Ace Glass
12192-32 12192-34 12192-36
76529-644EA 332.81 CAD
76529-644 76529-646 76529-648
Glass to Glass Gasket, NW Flange, Ace Glass
Seals Gaskets
NW (DN) flange glass to glass sealing gasket featuring a PTFE carrier and O-ring.
  • High temperature glass to glass seal
  • Made of PTFE
  • Excellent for vaccum work or a maximum of 2 bar pressure
Included Capfe O-ring is suitable for use in temperatures from –60 to +204 °C.