BASIC* Lab Coats, Halyard
Supplier: O&M HALYARD CANADA, INC CA
10120 10121 10122 10123 10124
CA76540-352CS 232.98 CAD
CA76540-352 CA76540-354 CA76540-356 CA76540-358 CA76540-360
BASIC* Lab Coats, Halyard
Disposable Garments
HALYARD* BASIC* Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
  • Traditional collar
  • Raglan sleeves
  • Knit cuffs
  • Four snap front closure
  • Two reinforced pockets
  • Size/name label
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application