PUREZERO* HG3 Light Blue Nitrile Gloves, Halyard
Supplier: O&M HALYARD CANADA, INC CA
CLN9031LG CLN9031MD CLN9031SM CLN9031XL CLN9031XS
CA76540-286CS 677.08 CAD
CA76540-286 CA76540-288 CA76540-290 CA76540-292 CA76540-294
PUREZERO* HG3 Light Blue Nitrile Gloves, Halyard
Gloves
PUREZERO* Cleanroom Gloves are ideal for applications that involve handling delicate equipment in microelectronics, semiconductors, optics, pharmaceutical, and medical device manufacturing applications. They are specifically designed to meet the stringent requirements of cleanroom environments.
  • Recommended for use in ISO Class 3 or higher and Grade A/B/C/D cleanrooms
  • Maximum particles: >0.5 µm/cm² less than 1200
  • AQL 0.65 for pinholes
  • Accelerator-, silicone-, and powder-free
  • 30.5 cm (12") length with beaded cuff
  • Fingertip thickness: 0.10 mm/3.9 mil
  • Palm
More Product Information