PUREZERO* HG3 White SGX* Nitrile Gloves, Halyard
Supplier: O&M HALYARD CANADA, INC CA
CLN3231LG CLN3231MD CLN3231SM CLN3231XL CLN3231XS
CA76540-260CS 519.34 CAD
CA76540-260 CA76540-262 CA76540-264 CA76540-266 CA76540-268
PUREZERO* HG3 White SGX* Nitrile Gloves, Halyard
Gloves
PUREZERO* Cleanroom Gloves are ideal for applications that involve handling delicate equipment in microelectronics, semiconductors, optics, pharmaceutical, and medical device manufacturing applications. They are specifically designed to meet the stringent requirements of cleanroom environments.
  • Recommended for use in ISO Class 3 or higher and Grade A/B/C/D cleanrooms
  • Maximum particles: >0.5 µm/cm² less than 950
  • AQL 0.65 for pinholes
  • Accelerator-, silicone-, latex-, and powder-free
  • 30.5 cm (12") length with beaded cuff
  • Fingertip thickness: 0.16 mm/6 mil
  • Palm thickness: 0.13 mm/5 mil
  • Smooth finish, static for dissipative use
These gloves are clean processed (washed repeatedly in deionized water) to ensure consistent control of low particles and extractables. They are recommended for use in ISO Class 3 or higher cleanroom environments. PUREZERO* HG3 Smooth White Nitrile Cleanroom Gloves are nonsterile, and ambidextrous with a smooth finish. They feature a beaded cuff with a 6 mil fingertip thickness and textured fingertips and palms. They have triple-layer cleanroom compatible packaging (double poly bagged with poly case liner) and are packaged in an ISO Class 5 cleanroom environment.
Because PUREZERO* HG3 cleanroom gloves are made with an accelerator-free nitrile polymer, they are an excellent alternative for workers with latex allergies.
Certifications: Certificates of Analysis (CoA), and Certificates of Irradiation (CoI) easily accessible online for every production lot. Declaration of Conformity (DoC) documents showing compliance to applicable regulations and standards are also available.